missing translation for 'onlineSavingsMsg'
Learn More

11-Heneicosanol, 97%, Thermo Scientific Chemicals

Product Code. 11341847
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3381-26-8
Molecular Formula C21H44O
Molecular Weight (g/mol) 312.582
MDL Number MFCD00026547
InChI Key BCNCKKYOXRHQGT-UHFFFAOYSA-N
Synonym 11-heneicosanol, 11-henicosanol, 11-henicosanol #, acmc-20aozg
PubChem CID 76913
IUPAC Name henicosan-11-ol
SMILES CCCCCCCCCCC(CCCCCCCCCC)O

Specifications

Melting Point 71°C to 73°C
Odor Odorless
Quantity 1 g
Beilstein 1765841
Formula Weight 312.58
Percent Purity 97%
Chemical Name or Material 11-Heneicosanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
11-Heneicosanol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.