Learn More
1S,3R-RSL3
Induces ferroptosis; inhibits glutathione peroxidase 4 (GPX4)
Marca: Tocris Bioscience 6118/50
Descripción
1S,3R-RSL3 is a glutathione peroxidase 4 (GPX4) inhibitor. Induces ferroptosis. Increases lipid peroxidase levels in vitro. Inhibits tumor formation and progression in vivo.Negative control 1R,3R-RSL3 (Cat. No. 6687) also available.
Especificaciones
(1S,3R)-Methyl 2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1H-pyrido[3,4-b]indole-3-carboxylate | |
>98% | |
C23H21ClN2O5 |
1219810-16-8 | |
50 mg | |
Observed MW: 440.88 |