missing translation for 'onlineSavingsMsg'
Learn More
2-Benzyloxy-2-methyl-1-propanol, 95%, Thermo Scientific Chemicals Product Code.: 15443337

2-Benzyloxy-2-methyl-1-propanol, 95%, Thermo Scientific Chemicals

Product Code. 15443337
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15443337

Brand: Thermo Scientific Alfa Aesar H26006.MD

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

2-Benzyloxy-2-methyl-1-propanol is used as a building block in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzyloxy-2-methyl-1-propanol is used as a building block in organic synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91968-71-7
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.25
MDL Number MFCD08459314
InChI Key FRHJVAJQSPVSKV-UHFFFAOYSA-N
Synonym 2-benzyloxy-2-methylpropan-1-ol, 2-benzyloxy-2-methyl-1-propanol, 2-methyl-2-phenylmethoxy propan-1-ol
PubChem CID 42552903
IUPAC Name 2-methyl-2-phenylmethoxypropan-1-ol
SMILES CC(C)(CO)OCC1=CC=CC=C1

Specifications

Boiling Point 80°C to 85°C (0.1 mmHg)
Quantity 250 mg
Solubility Information Immiscible with water.
Formula Weight 180.12
Percent Purity 95%
Chemical Name or Material 2-Benzyloxy-2-methyl-1-propanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10143658 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26006.03,H26006.MD
Lot NumberI29T001 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26006.03,H26006.MD
Lot Number10229594 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26006.03,H26006.MD
Lot Number10140124 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26006.03,H26006.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Benzyloxy-2-methyl-1-propanol, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.