missing translation for 'onlineSavingsMsg'
Learn More
Alfa Aesar™ 2-Benzyloxyaniline, 97% Product Code.: 11446120

Alfa Aesar™ 2-Benzyloxyaniline, 97%

Product Code. 11446120
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11446120

Brand: Alfa Aesar™ H27065.06

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 20012-63-9
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.253
MDL Number MFCD03093873
InChI Key PLPVLSBYYOWFKM-UHFFFAOYSA-N
Synonym 2-benzyloxyaniline, 2-benzyloxy aniline, benzenamine, 2-phenylmethoxy, 2-benzyloxyanilline, 2-benzyloxy-aniline, acmc-1cmch, 2-aminophenyl benzyl ether, 3-amino-4-benzyloxybenzene
PubChem CID 240548
IUPAC Name 2-phenylmethoxyaniline
SMILES C1=CC=C(C=C1)COC2=CC=CC=C2N

Specifications

Melting Point 35°C to 39°C
Quantity 5 g
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material 2-Benzyloxyaniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Alfa Aesar™ 2-Benzyloxyaniline, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.