missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-3,5-bis(trifluoromethyl)aniline, 98%, Thermo Scientific™ Product Code.: 15411179

2-Chloro-3,5-bis(trifluoromethyl)aniline, 98%, Thermo Scientific™

Product Code. 15411179
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15411179

Brand: Thermo Scientific Alfa Aesar H64119.22

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-3,5-bis(trifluoromethyl)aniline is used as dyestuff Intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201593-90-0
Molecular Formula C8H4ClF6N
Molecular Weight (g/mol) 263.567
MDL Number MFCD04038716
InChI Key JKUFETFGEJPHEM-UHFFFAOYSA-N
Synonym 2-chloro-3,5-bis trifluoromethyl aniline, 3,5-bis trifluoromethyl-2-chloroaniline, 2-chloro-3,5-bis-trifluoromethyl aniline, benzenamine, 2-chloro-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-2-chlorophenylamine, pubchem2938, 2-chloro-3,5-bis-trifluoromethyl-aniline, 3,5-bis trifluoromethyl-2-chloro aniline, 2-chloranyl-3,5-bis trifluoromethyl aniline, 2-chloro-3,5-bis-trifluoromethyl-phenylamine
PubChem CID 2757429
IUPAC Name 2-chloro-3,5-bis(trifluoromethyl)aniline
SMILES C1=C(C=C(C(=C1C(F)(F)F)Cl)N)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.595 g/mL
Refractive Index 1.459
Quantity 100 g
UN Number UN2810
Beilstein 13730810
Solubility Information Slightly soluble in water.
Formula Weight 263.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloro-3,5-bis(trifluoromethyl)aniline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81302318 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-3,5-bis(trifluoromethyl)aniline, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.