missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals 2'-Deoxyuridine-5'-monophosphate disodium salt, 99% Product Code.: 15480377

Thermo Scientific Chemicals 2'-Deoxyuridine-5'-monophosphate disodium salt, 99%

Product Code. 15480377
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15480377

Brand: Thermo Scientific Chemicals J64627.03

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

2?-Deoxyuridine 5?-monophosphate (dUMP) is used by the enzyme thymidylate synthetase (TS) (EC 2.1.1.45) for the de novo production of dTMP. dUMP is used as the reference substrate in studies of dUMP analogues in potential chemotherapy applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyuridine 5′-monophosphate (dUMP) is used by the enzyme thymidylate synthetase (TS) (EC 2.1.1.45) for the de novo production of dTMP. dUMP is used as the reference substrate in studies of dUMP analogues in potential chemotherapy applications.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42155-08-8
Molecular Formula C9H13N2Na2O8P
Molecular Weight (g/mol) 354.162
MDL Number MFCD00065282
InChI Key CWTUMDATELIIAI-CDNBRZBRSA-N
Synonym 2'-deoxyuridine 5'-monophosphate disodium salt
PubChem CID 129893468
IUPAC Name [(2R,3S,5R)-5-(2,4-dioxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
SMILES C1C(C(OC1N2C=CC(=O)NC2=O)COP(=O)(O)O)O.[Na].[Na]

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 352.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Deoxyuridine-5'-monophosphate disodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX01K019 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog NumberJ64627.03,J64627.MD
Lot NumberN19K030 Certificate TypeCertificate of Analysis Date06/03/2024 Catalog NumberJ64627.03,J64627.MD
Lot NumberM11H091 Certificate TypeCertificate of Analysis Date07/02/2024 Catalog NumberJ64627.03,J64627.MD
Lot NumberN16J008 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberJ64627.03
Lot NumberY15I036 Certificate TypeCertificate of Analysis Date29/11/2022 Catalog NumberJ64627.03,J64627.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 2'-Deoxyuridine-5'-monophosphate disodium salt, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.