missing translation for 'onlineSavingsMsg'
Learn More
3-Amino-4-[(1-benzyl-2-methoxy-2-oxoethyl)amino]-4-oxobutanoic acid, 97%, Thermo Scientific™ Product Code.: 10476862

3-Amino-4-[(1-benzyl-2-methoxy-2-oxoethyl)amino]-4-oxobutanoic acid, 97%, Thermo Scientific™

Product Code. 10476862
10 g
Click to view available options
Quantity:
10 g
50 g
100 g
This item is not returnable. View return policy

Product Code. 10476862

Brand: Thermo Scientific Maybridge SB00761EA

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Chemical Identifiers

CAS 25548-16-7
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.31
MDL Number MFCD00176858
InChI Key IAOZJIPTCAWIRG-UHFFFAOYNA-N
Synonym aminosweet, equal, tri-sweet, 3-amino-4-1-benzyl-2-methoxy-2-oxoethyl amino-4-oxobutanoic acid, canderel, sweet dipeptide, aspartame usan:ban:inn, methyl alpha-aspartylphenylalaninate, phenylalanine,a-aspartyl-, 2-methyl ester, sanecta
PubChem CID 2242
SMILES COC(=O)C(CC1=CC=CC=C1)NC(=O)C(N)CC(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
IUPAC Name 3-amino-3-[(1-methoxy-1-oxo-3-phenylpropan-2-yl)carbamoyl]propanoic acid
Formula Weight 294.31
Percent Purity 97%
Chemical Name or Material 3-Amino-4-[(1-benzyl-2-methoxy-2-oxoethyl)amino]-4-oxobutanoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Amino-4-[(1-benzyl-2-methoxy-2-oxoethyl)amino]-4-oxobutanoic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.