missing translation for 'onlineSavingsMsg'
Learn More
4-Chloro-3-(trifluoromethylthio)benzyl bromide, 97%, Thermo Scientific™ Product Code.: 11326709

4-Chloro-3-(trifluoromethylthio)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11326709
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11326709

Brand: Thermo Scientific Alfa Aesar H26246.03

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886503-38-4
Molecular Formula C8H5BrClF3S
Molecular Weight (g/mol) 305.537
MDL Number MFCD06660314
InChI Key QRIMMRPSWRLACM-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethylthio benzyl bromide, 4-bromomethyl-1-chloro-2-trifluoromethyl sulfanyl benzene
PubChem CID 17750740
IUPAC Name 4-(bromomethyl)-1-chloro-2-(trifluoromethylsulfanyl)benzene
SMILES C1=CC(=C(C=C1CBr)SC(F)(F)F)Cl

Specifications

Refractive Index 1.544
Quantity 1 g
UN Number UN3265
Formula Weight 305.54
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethylthio)benzyl bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10141534 Certificate TypeCertificate of Analysis Date02/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-3-(trifluoromethylthio)benzyl bromide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.