missing translation for 'onlineSavingsMsg'
Learn More
4-Fluoro-3-methyl-5-(trifluoromethyl)cinnamic acid, 97%, Thermo Scientific™ Product Code.: 15418615

4-Fluoro-3-methyl-5-(trifluoromethyl)cinnamic acid, 97%, Thermo Scientific™

Product Code. 15418615
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15418615

Brand: Thermo Scientific Alfa Aesar H33725.MD

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1373921-81-3
Molecular Formula C11H8F4O2
Molecular Weight (g/mol) 248.177
MDL Number MFCD22201075
InChI Key JWTWWSLPXHUYNI-NSCUHMNNSA-N
Synonym 4-fluoro-3-methyl-5-trifluoromethyl cinnamic acid, 2e-3-4-fluoro-3-methyl-5-trifluoromethyl phenyl prop-2-enoic acid, 3-4-fluoro-3-methyl-5-trifluoromethyl phenyl acrylic acidShow MoreShow Less
PubChem CID 86277682
IUPAC Name (E)-3-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]prop-2-enoic acid
SMILES CC1=C(C(=CC(=C1)C=CC(=O)O)C(F)(F)F)F

Specifications

Quantity 250 mg
Formula Weight 248.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methyl-5-(trifluoromethyl)cinnamic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10178049 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Fluoro-3-methyl-5-(trifluoromethyl)cinnamic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.