missing translation for 'onlineSavingsMsg'
Learn More

4-Picoline, 98%, Thermo Scientific Chemicals

Product Code. 11487447
500 mL
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.

Solubility
Soluble in water, alcohol and ether.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, water & moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-89-4
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD00006440
InChI Key FKNQCJSGGFJEIZ-UHFFFAOYSA-N
Synonym 4-picoline, pyridine, 4-methyl, p-picoline, gamma-picoline, p-methylpyridine, 4-methyl-pyridine, gamma-methylpyridine, unii-tjd6v9sso7, .gamma.-picoline, 4-mepy
PubChem CID 7963
ChEBI CHEBI:32547
IUPAC Name 4-methylpyridine
SMILES CC1=CC=NC=C1

Specifications

Melting Point 3°C
Density 0.957
pH 10 to 11
Boiling Point 144°C to 145°C
Flash Point 39°C (102°F)
Odor Unpleasant
Refractive Index 1.505
Quantity 500 mL
UN Number UN2313
Beilstein 104586
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,7402
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 93.13
Percent Purity 98%
Chemical Name or Material 4-Picoline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Picoline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.