missing translation for 'onlineSavingsMsg'
Learn More
4-(Trifluoromethylthio)benzyl alcohol, 97%, Thermo Scientific™ Product Code.: 11426946

4-(Trifluoromethylthio)benzyl alcohol, 97%, Thermo Scientific™

Product Code. 11426946
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 11426946

Brand: Thermo Scientific Alfa Aesar B24026.03

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56456-52-1
Melting Point 49°C to 54°C
Molecular Formula C8H7F3OS
MDL Number MFCD00236342
Quantity 1 g
Beilstein 8616292
Synonym 4-trifluoromethylthio benzyl alcohol, 4-trifluoromethyl thio phenyl methanol, 4-trifluoromethylsulfanyl phenyl methanol, 4-trifluoromethyl sulfanyl phenyl methanol, 4-trifluoromethylthiobenzyl alcohol, 4-trifluoromethylthio benzylalcohol, 4-trifluoromethylsulfanyl-phenyl-methanol, 4-trifluoromethylthio phenyl methanol, 4-trifluoromethylthio phenyl methan-1-ol
InChI Key KCWQNXIWNNTUIJ-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CO)SC(F)(F)F
IUPAC Name [4-(trifluoromethylsulfanyl)phenyl]methanol
Molecular Weight (g/mol) 208.198
PubChem CID 2777870
Formula Weight 208.2
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethylthio)benzyl alcohol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10235821 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberJ27W022 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10171657 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Trifluoromethylthio)benzyl alcohol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.