missing translation for 'onlineSavingsMsg'
Learn More
5-(2-Methyl-1,3-thiazol-4-yl)-2-thiophenecarboxylic acid, 97%, Thermo Scientific™ Product Code.: 10587844

5-(2-Methyl-1,3-thiazol-4-yl)-2-thiophenecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10587844
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10587844

Brand: Thermo Scientific Maybridge CC18201DE

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Chemical Identifiers

CAS 400715-45-9
Molecular Formula C9H7NO2S2
Molecular Weight (g/mol) 225.28
MDL Number MFCD02682053
InChI Key AEDWMWQOFJYAGN-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid, 2-thiophenecarboxylicacid, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-carboxylic acid, 5-2-methylthiazol-4-yl-thiophene-2-carboxylic acid, 5-2-methylthiazol4-yl-thiophene-2-carboxylic acid, thiophene-2-carboxylic acid, 5-2-methyl-4-thiazolyl, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid #
PubChem CID 614467
IUPAC Name 5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carboxylic acid
SMILES CC1=NC(=CS1)C2=CC=C(S2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 225.29
Percent Purity 97%
Chemical Name or Material 5-(2-Methyl-1, 3-thiazol-4-yl)-2-thiophenecarboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-(2-Methyl-1,3-thiazol-4-yl)-2-thiophenecarboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.