Learn More
CD45RA Mouse anti-Human, APC-H7, Clone: HI100, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 560674
285.07 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Descripción
The HI100 monoclonal antibody specifically binds to the 220 kDa isoform of the human leukocyte common antigen, CD45RA. CD45RA is expressed on approximately 40-50% of peripheral CD4+ T cells, 50% of peripheral CD8+ T cells and on a portion of B cells and monocytes. The CD45RA antigen is expressed by naοve and activated T cells. CD45RA-specific antibodies are useful for the study of the suppressor/inducer subpopulation of CD4+ lymphocytes.

Especificaciones
CD45RA | |
Monoclonal | |
APC-H7 | |
Mouse | |
50 Tests | |
Immunology | |
Human | |
IgG2b κ |
Flow Cytometry | |
HI100 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Sugerencias de productos
Customers who viewed this item also viewed.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.