Learn More
CD8 Mouse anti-Human, PE-Cy7, Clone: RPA-T8, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 557750
367.44 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Descripción
Clone RPA-T8 reacts with the α subunit (32kDa) of the two-chain complex expressed on T-cytoxic/suppressor cell populations. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. RPA-T8 reacts with approximately 13-48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. RPA-T8 and HIT8a (Mfr. No.555337) are not cross-blocking. This clone also reacts with a subset of peripheral blood lymphocytes, but not monocytes nor granuloyctes, of baboon and both rhesus and cynomolgus macaque monkey. In general, a higher frequency of CD8+ and CD4+CD8+ lymphocytes are observed in non-human primates compared to normal human donors. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Especificaciones
CD8 | |
Monoclonal | |
PE-Cyanine7 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human CD8a | |
50 Tests | |
Immunology | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
RPA-T8 | |
CD8α; CD8A; CD8 alpha; Leu2; MAL; T8; p32 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Sugerencias de productos
Customers who viewed this item also viewed.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.