missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ral GEF with PH domain and SH3 binding motif 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human RALGPS2 protein.
Marca: Abnova H00055103-D01P.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Sequence: MDLMNGQASSVNIAATASEKSSSSESLSDKGSELKKSFDAVVFDVLKVTPEEYAGQITLMDVPVFKAIQPDELSSCGWNKKEKYSSAPNAVAFTRRFNHVSFWVVREILHAQTLKIRAEVLSHYIKTAKKLYELNNLHALMAVVSGLQSAPIFRLTKTWALLSRKDKTTFEKLEYVMSKEDNYKRLRDYISSLKMTPCIPYLGIYLSDLTYIDSAYPSTGSILENEQRSNLMNNILRIISDLQQSCEYDIPMLPHVQKYLNSVQYIEELQKFVEDDNYKEspecificaciones
Ral GEF with PH domain and SH3 binding motif 2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human RALGPS2 protein. | |
RALGPS2 | |
FLJ10244/FLJ25604/KIAA0351/dJ595C2.1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_018037 | |
RALGPS2 | |
RALGPS2 (NP_060507.1, 1 a.a. ∼ 279 a.a) full-length human protein. | |
100 μg | |
Signal Transduction | |
55103 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Ral GEF with PH domain and SH3 binding motif 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™ > 100μg; Unlabeled
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido