missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ataxin 7 Monoclonal Antibody (3SCA-1C1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA3083
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA3-083 detects Ataxin 7 from human and mouse samples. MA3-083 has been successfully used in Western blot, immunofluorescence, and ELISA applications. The MA3-083 epitope is mapped between aa 66 and aa 119.
Spinocerebellar ataxia type 7 (SCA7) belongs to a class of hereditary neurodegenrative diseases characterized by abnormally expanded polyglutamine (CAG) repeats at the N-terminus. This autosomal dominant cerebellar ataxia primarily affects the cerebellum, retina, and brain stem and causes dementia, macular degeneration and other neurodegenerative characteristics. SCA7 gene encodes the ubiquitous 892 residue protein ataxin-7. This protein is typically located in the cytoplasm and on the nuclear membrane of normal brain neurons. In cells where there is a mutation of the SCA7 gene, ataxin 7 accumulates in intranuclear inclusions and can result in cell death.
Especificaciones
Ataxin 7 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.05% sodium azide | |
O15265, Q8R4I1 | |
ATXN7 | |
Recombinant human Ataxin 7 aa 1 - 229. | |
RUO | |
246103, 6314 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
3SCA-1C1 | |
Unconjugated | |
ATXN7 | |
A430107N12Rik; ADCAII; AI627028; Ataxin 7; Ataxin7; Ataxin-7; ATXN7; Atxn7-PA; Atxn7-PB; Atxn7-PC; Atxn7-PD; CG9866; CG9866-PA; CG9866-PB; CG9866-PC; CG9866-PD; Dmel\CG9866; Dmel_CG9866; LD40170p; OPCA3; RGD1562692; SCA7; spinocerebellar ataxia 7 homolog; spinocerebellar ataxia type 7 protein; Spinocerebellar ataxia type 7 protein homolog | |
Mouse | |
50 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Ataxin 7 Monoclonal Antibody (3SCA-1C1), Invitrogen™ > 50 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido