missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AXIN1 Monoclonal Antibody (T.618.2), Invitrogen™
Rabbit Monoclonal Antibody
Marca: Invitrogen MA514853
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
It is not recommended to aliquot this antibody.
AXIN1 is a cytoplasmic protein which contains a regulation of G-protein signaling (RGS) domain and a disheveled and axin (DIX) domain and is thought to function as a negative regulator of the WNT signaling pathway that regulates embryonic axis formation. AXIN1 interacts with adenomatosis polyposis coli (APC), beta-catenin, glycogen synthase kinase 3 beta, forming a tetrameric complex resulting in the regulation of the stabilization of beta-catenin. Mutations in the AXIN1 gene have been associated various carcinomas, indicating that it also functions as a tumor suppressor.
Especificaciones
AXIN1 | |
Monoclonal | |
140 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
O15169, O35625 | |
AXIN1 | |
Recombinant protein of human Axin1 | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
T.618.2 | |
Unconjugated | |
AXIN1 | |
AI316800; AXIN; axin 1; Axin1; axin-1; Axis inhibition protein 1; axis inhibitor 1; Fu; fused; fused, mouse, homolog of; GSK-3beta interacting protein rAxin; hAxin; Kb; Ki; kinky; knobbly; LA16c-314G4.3; MGC52315; PPP1R49; Protein Fused; protein phosphatase 1, regulatory subunit 49; rAxin | |
Rabbit | |
Affinity chromatography | |
RUO | |
12005, 8312 | |
-20°C | |
Liquid |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
AXIN1 Monoclonal Antibody (T.618.2), Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido