missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BECN1 Mouse anti-Canine, Human, Rat, Clone: OTI1F1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF502643

This item is not returnable.
View return policy
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Beclin 1 plays a central role in autophagy. It acts as a core subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate. Different PI3K complex forms are believed to play a role in multiple membrane trafficking pathways - PI3KC3-C1 is involved in the initation of autophagosomes and PI3KC3-C2 is involved in the maturation of autophagosomes and endocytosis. Beclin 1 also plays a role in antiviral host defense.Specifications
BECN1 | |
Monoclonal | |
Unconjugated | |
BECN1 | |
ATG6, VPS30, beclin1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
114558, 480513, 8678 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1F1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q14457, Q91XJ1 | |
Becn1 | |
Full length human recombit protein of human BECN1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG2a |
Product Suggestions
Los clientes que vieron este artículo también vieron
Viewing 1-4 of
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
BECN1 Mouse anti-Canine, Human, Rat, Clone: OTI1F1, lyophilized, TrueMAB™
Spot an opportunity for improvement?Share a Content Correction