missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blood Group Lewis A Monoclonal Antibody (7LE), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA119344
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
For IHC (P), heat mediated antigen retrieval in sodium citrate is recommended.
The Lewis histo-blood group system comprises a set of fucosylated glycosphingolipids that are synthesized by exocrine epithelial cells and circulate in body fluids. The glycosphingolipids function in embryogenesis, tissue differentiation, tumor metastasis, inflammation, and bacterial adhesion. They are secondarily absorbed to red blood cells giving rise to their Lewis phenotype. This gene is a member of the fucosyltransferase family, which catalyzes the addition of fucose to precursor polysaccharides in the last step of Lewis antigen biosynthesis. It encodes an enzyme with alpha(1,3)-fucosyltransferase and alpha(1,4)-fucosyltransferase activities. Mutations in this gene are responsible for the majority of Lewis antigen-negative phenotypes. Multiple alternatively spliced variants, encoding the same protein, have been found for this gene.
Especificaciones
Blood Group Lewis A | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P21217 | |
FUT3 | |
Mucin from ovarian cyst fluid. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
7LE | |
Unconjugated | |
FUT3 | |
4-fucosyltransferase; alpha-(1,3/1,4)-fucosyltransferase; blood group Lewis alpha-4-fucosyltransferase; CD174; FT3B; Fucosyltransferase 3; fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood group); fucosyltransferase 3 (Lewis blood group); fucosyltransferase III; FucT-III; FUT3; Galactoside 3(4)-L-fucosyltransferase; LE; Les; Lewis FT; Lewis Y | |
Mouse | |
Protein A | |
RUO | |
2525 | |
4° C, do not freeze | |
Liquid |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Blood Group Lewis A Monoclonal Antibody (7LE), Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido