Learn More
Capreomycin sulfate, MedChemExpress
MedChemExpress Capreomycin sulfate is a peptide antibiotic, commonly grouped with the aminoglycosides, which is given in combination with other antibiotics for MDR-tuberculosis.
238.95€ - 637.20€
Especificaciones
Nombre del producto químico o material | Capreomycin sulfate |
---|---|
CAS | 1405-37-4 |
Color | White |
Notas de grado de pureza | Research |
Fórmula molecular | C25H46N14O11S |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16499867
|
Medchem Express
HY-17566 |
5 g |
637.20€
5 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16451699
|
Medchem Express
HY-17566 |
1 g |
238.95€
1 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
Capreomycin sulfate is a peptide antibiotic, commonly grouped with the aminoglycosides, which is given in combination with other antibiotics for MDR-tuberculosis.Specifications
Capreomycin sulfate | |
White | |
C25H46N14O11S | |
O=C(CC(N)CCCN)NCC(C(N/C(C(NC(C1NC(NCC1)=N)C2=O)=O)=C/NC(N)=O)=O)NC(C(NC(C(CN2)N)=O)C)=O.O=S(O)(O)=O | |
750.78 | |
Research | |
COVID-19-immunoregulation |
1405-37-4 | |
Research | |
H2O : ≥ 37 mg/mL (49.28 mM) | |
750.78 | |
98.07% | |
Powder |
Safety and Handling
H302∣H315∣H319∣H332∣H335
missing translation for 'shelfLife' : -20°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : -20°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.