missing translation for 'onlineSavingsMsg'
Learn More

CD90.1 Mouse, PE, Clone: OX-7, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 551401

346.62 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.


 Ver más versiones de este producto

Código de producto. 15896638

  • 347.00€ / cada uno

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

CD90 (Thy-1) is a GPI-anchored membrane glycoprotein of the Ig superfamily which is involved in signal transduction. The OX-7 clone reacts with rat CD90 reported to be expressed by hematopoietic stem cells, early myeloid & erythroid cells, immature B lymphocytes in the bone marrow & peripheral lymphoid organs, thymocytes, recent thymic emigrants (a subset of CD45RC- peripheral T lymphocytes), neurons, glomerular mesangial cells, endothelium at inflammatory sites, mast cells, and dendritic cells. Rat dendritic epidermal T cells (DEC) have been
reported to be CD90 (Thy-1) negative, unlike those of the mouse. The OX-7 clone has been reported to crossreact with the mouse CD90.1 (Thy-1.1) alloantigen of the AKR/J and PL strains, but not CD90.2 (Thy-1.2) found on many mouse strains. In the mouse, CD90 is found on thymocytes, most peripheral T lymphocytes, some intraepithelial T lymphocytes (IEL, DEC), hematopoietic stem cells, and neurons, but not B lymphocytes. In addition, there is evidence that CD90 mediates adhesion of mouse thymocytes to mouse thymic stroma. The OX-7 clone has also been reported to crossreact with rabbit and guinea pig thymus, brain, and intestine. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: OX-7
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Rat thymocyte Thy-1 antigen

Flow Cytometry

TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

CD90.1
Monoclonal
0.2mg/mL
Rat Thy-1; Mouse Thy-1.1
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
OX-7
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat thymocyte Thy-1 antigen
0.1 mg
Immunology
Guinea Pig, Mouse, Rabbit, Rat
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
CD90.1 Mouse, PE, Clone: OX-7, BD > 0.1mg; PE

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado

For Research Use Only.