missing translation for 'onlineSavingsMsg'
Learn More
Cyclopentadienylmolybdenum(V) tetrachloride, 95%, Thermo Scientific Chemicals Product Code.: 11308518

Cyclopentadienylmolybdenum(V) tetrachloride, 95%, Thermo Scientific Chemicals

Product Code. 11308518
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11308518

Brand: Thermo Scientific Alfa Aesar 040522.03

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62927-99-5
MDL Number MFCD00269826
Synonym Tetrachlorocyclopentadienylmolybdenum(V)

Specifications

Color Purple-Red
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 302.85
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Cyclopentadienylmolybdenum(V) tetrachloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP03I799 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number040477.M1,040477.K2,040477.M6,040477.K7,040522.03
Lot NumberJ15Y912 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number040477.M1,040477.K2,040477.M6,040477.K7,040522.03
Lot NumberJ15W807 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number040477.M1,040477.K2,040477.M6,040477.K7,040522.03
Lot NumberJ15R805 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number040477.M1,040477.K2,040477.M6,040477.K7,040522.03
Lot NumberJ05U906 Certificate TypeCertificate of Analysis Date29/07/2022 Catalog Number040477.M1,040477.K2,040477.M6,040477.K7,040522.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclopentadienylmolybdenum(V) tetrachloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.