missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP3 Monoclonal Antibody (2#), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA543847
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Protein tyrosine phosphatases (PTPs) are a group of enzymes that remove phosphate groups from phosphorylated tyrosine residues on proteins. Together with tyrosine kinases, PTPs regulate the phosphorylation state of many important signaling molecules, such as the MAP kinase family. Recently, increasing attention has been focused on the growing family of PTPs. Like PTKs, PTPs have been implicated in cell signaling, cell growth and proliferation, and oncogenic transformation. Moreover, some PTPs can be involved in cell cycle regulation and embryogenesis. Dual specificity phosphatases (DSPs) are an emerging subclass of the protein tyrosine phosphatase (PTP) gene superfamily, which appears to be selective for dephosphorylating the critical phosphothreonine and phosphotyrosine residues. The prototypical DSP is the VH1 gene in vaccinia virus expressed in late-stage viral infection. A shallow active site pocket in VHR allows for the hydrolysis of phosphorylated serine, threonine, or tyrosine protein residues, whereas the deeper active site of protein tyrosine phosphatases (PTPs) restricts substrate specificity to only phosphotyrosine.
Especificaciones
DUSP3 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P51452 | |
Dusp3 | |
Ser2-Pro185 (Accession P51452), with N-terminal His Tag | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG2a κ |
Immunohistochemistry (Paraffin), Western Blot | |
2# | |
Unconjugated | |
Dusp3 | |
2210015O03Rik; 5031436O03Rik; dual specificity phosphatase 3; dual specificity phosphatase 3 (vaccinia virus phosphatase VH1-related); dual specificity phosphatase 3 (vaccinia virus phosphatase VH1-related)-like; dual specificity protein phosphatase 3; Dual specificity protein phosphatase VHR; DUSP 3; DUSP3; DUSP-3; RGD1560049; serine/threonine specific protein phosphatase; T-DSP11; Vaccinia H1-related phosphatase; vaccinia virus phosphatase VH1-related; VHR | |
Mouse | |
Protein A/G | |
RUO | |
1845, 498003 | |
-20°C | |
Liquid |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
DUSP3 Monoclonal Antibody (2#), Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido