missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 2-methyl-1,5-diphenylpyrrole-3-carboxylate, 97%, Thermo Scientific™ Product Code.: 11478677

Ethyl 2-methyl-1,5-diphenylpyrrole-3-carboxylate, 97%, Thermo Scientific™

Product Code. 11478677
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11478677

Brand: Thermo Scientific Alfa Aesar A19832.03

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3652-61-7
Molecular Formula C20H19NO2
Molecular Weight (g/mol) 305.377
MDL Number MFCD00221065
InChI Key YHEGUULWFOVVRG-UHFFFAOYSA-N
Synonym ethyl 2-methyl-1,5-diphenyl-1h-pyrrole-3-carboxylate, acmc-20al0y, maybridge1_008409, 1,5-diphenyl-2-methyl-1h-pyrrole-3-carboxylic acid ethyl ester, 1h-pyrrole-3-carboxylicacid, 2-methyl-1,5-diphenyl-, ethyl ester
PubChem CID 2742174
IUPAC Name ethyl 2-methyl-1,5-diphenylpyrrole-3-carboxylate
SMILES CCOC(=O)C1=C(N(C(=C1)C2=CC=CC=C2)C3=CC=CC=C3)C

Specifications

Melting Point 95°C to 99°C
Odor Odorless
Quantity 1 g
Formula Weight 305.38
Percent Purity 97%
Chemical Name or Material Ethyl 2-methyl-1,5-diphenylpyrrole-3-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 2-methyl-1,5-diphenylpyrrole-3-carboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.