Learn More
GAB1 & SOS1 Protein Protein Interaction Antibody Pair, Abnova™
Antibody Pair
Marca: Abnova DI0185.1Set
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the GAB1 protein, and the other against the SOS1 protein for use in in situ Proximity Ligation Assay.Especificaciones
GAB1 | |
Unconjugated | |
- | |
1 Set | |
Primary | |
Antibody pair set content: 1. GAB1 rabbit purified polyclonal antibody (20 ug) 2. SOS1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
In situ PLA | |
GRB2-associated binding protein 1 | |
GAB1 | |
RUO | |
2549/6654 | |
Human | |
Ab Pair |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-5 of 12
Título del producto
GAB1 & SOS1 Protein Protein Interaction Antibody Pair, Abnova™ > 1 Set; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido