missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Armenian Hamster IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 647

Goat Polyclonal Secondary Antibody

Marca:  Invitrogen™ A78967

276.45 EUR hasta el 2025-05-31
Use el código promocional "25977" para aplicar la oferta.


Código de producto. 17877071

  • 297.00€ / 500 microgramos

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

This antibody specifically detects Armenian hamster IgG and shows negligible reactivity to Syrian hamster IgG as tested by ELISA and immunofluorescence. This goat anti-armenian hamster IgG (H+L) whole secondary antibody has been affinity-purified and shows minimum cross-reactivity to bovine, human, mouse, rabbit, rat serum proteins. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondary antibodies flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g. flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Invitrogen™ Alexa Fluor 647 dye is a near-infrared-fluorescent dye with excitation ideally suited to the 647 nm laser line. For stable signal generation in imaging and flow cytometry, Alexa Fluor 647 dye is pH-insensitive over a wide molar range. Probes with high fluorescence quantum yield and high photostability allow detection of...

Anti-Hamster secondary antibodies are affinity-purified antibodies with well-characterized specificity for hamster immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

Armenian Hamster IgG (H+L) Highly Cross-Adsorbed
Polyclonal
Alexa Fluor 647
Goat
purified
RUO
Hamster
Secondary Antibody
IgG
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
2 mg/mL
PBS with 5mM sodium azide; pH 7.5
Gamma Immunoglobins Heavy and Light chains.
500 μg
Secondary
4°C, store in dark, DO NOT FREEZE!
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

  • Multiple Manufacturers
    Desbloquee avances en proteómica y ahorre de manera exclusiva.
    Promo Code: 25977

    Expires2025-05-31

    Days

    44

    Hours

    18

    Minutes

    47

    Seconds

    10
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Invitrogen™ Goat anti-Armenian Hamster IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 647 > 500 μg; Alexa Fluor 647

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado