Learn More
O&M Halyard Sequential Sterilization Wrap (H500)
Halyard Sequential wraps are the industry standard for sterile processing teams.
Marca: O&M 68145
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Tear-, flame-, lint- and abrasion-resistant
- POWERGUARD technology for enhanced microbial barrier protection
- Pre-vacuum steam sterilant penetration
- Ethylene Oxide (EO) sterilant penetration and residuals
- Maintenance of package integrity (MPI) to meet FDA requirements
- Instructions for Use that meet FDA guidelines
- Class 1 flammability rating
- Low linting
- Made in the USA
Especificaciones
Sterilization Wrap | |
Meets NFPA Class 1 | |
Heavy instrument sets, retractors, and applications where frequent handling occurs (13 to 17 lb.) | |
No estéril | |
114 x 114 cm (45 x 45 in.) | |
114 cm | |
114 cm |
Autoclavable | |
Azul | |
SMS Fabric (H500) | |
Sí | |
45 in. | |
45 in. |
Título del producto
O&M Halyard Sequential Sterilization Wrap (H500) > H500; 114 x 114 cm (45 x 45 in.)
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido