missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat GAPDH Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 11 publications
Marca: R&D Systems MAB5718
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
GAPDH Monoclonal specifically detects GAPDH in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Especificaciones
GAPDH | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, G3PDH, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2597 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
686613 | |
Western Blot 0.05 ug/mL, Simple Western 0.25 - 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P04406 | |
GAPDH | |
E. coli-derived recombinant human GAPDH/G3PDH Met1-Ala150 Accession # P04406 | |
100 μg | |
Primary | |
Detects human, mouse and rat GAPDH/G3PDH in Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse GAPDH/G3PDH and no cross-reactivity with recombinant human GAPDH-2 is observed. | |
Human, Mouse, Rat | |
Purified |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Human/Mouse/Rat GAPDH Antibody, R&D Systems™ > 100ug; Unlabeled
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido