missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNK Mouse anti-Human, Mouse, Rat, Clone: 1A12E1, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Especificaciones
Antígeno | JNK |
---|---|
Clon | 1A12E1 |
Concentración | 0.8 mg/mL |
Aplicaciones | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Clasificación | Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16867875
|
Proteintech
66210-1-IG-20UL |
20 μL |
145.00€
20 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16857875
|
Proteintech
66210-1-IG-150UL |
150 μL |
410.00€
150 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
This protein can be phosphorylated and this antibody recognizes the 46 kDa and 55 kDa bands in western blot.
C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predomit forms are JNK1 and JNK2.Especificaciones
JNK | |
0.8 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
c Jun N terminal kinase 1, JNK, JNK 46, JNK1, JNK1A2, JNK21B1/2, MAP kinase 8, MAPK 8, MAPK8, PRKM8, SAPK1 | |
MAPK10, MAPK8, Mapk9 | |
IgG1 | |
Protein G | |
Antibody |
1A12E1 | |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P45983, P45984, P49185, P49186, P49187, P53779, Q61831, Q91Y86, Q9WTU6 | |
116554, 25272, 26414, 26419, 26420, 50658, 5599, 5601, 5602 | |
JNK Fusion Protein Ag21426 | |
Primary | |
-20°C | |
MAPK10 |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
JNK Mouse anti-Human, Mouse, Rat, Clone: 1A12E1, Proteintech