missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNK Mouse anti-Human, Mouse, Rat, Clone: 1A12E1, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66210-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This protein can be phosphorylated and this antibody recognizes the 46 kDa and 55 kDa bands in western blot.
C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predomit forms are JNK1 and JNK2.Especificaciones
JNK | |
Monoclonal | |
0.8 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P45983, P45984, P49185, P49186, P49187, P53779, Q61831, Q91Y86, Q9WTU6 | |
MAPK10, MAPK8, Mapk9 | |
JNK Fusion Protein Ag21426 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1A12E1 | |
Unconjugated | |
MAPK10 | |
c Jun N terminal kinase 1, JNK, JNK 46, JNK1, JNK1A2, JNK21B1/2, MAP kinase 8, MAPK 8, MAPK8, PRKM8, SAPK1 | |
Mouse | |
Protein G | |
RUO | |
116554, 25272, 26414, 26419, 26420, 50658, 5599, 5601, 5602 | |
-20°C | |
Liquid |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
JNK Mouse anti-Human, Mouse, Rat, Clone: 1A12E1, Proteintech > 150 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido