Learn More
Eurofins DiscoverX Lck Protein, Active
Active, N-terminal His6-tagged, recombinant full-length human Lck, for use in Kinase Assays.
525.15€ - 4008.15€
Especificaciones
Formulación | mg/mL of enzyme in 50 mM Tris/HCl, pH 7.5, 0.1 mM EGTA, 150 mM NaCl, 1 mM benzamidine, 0.03% Brij-35, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 5% glycerol. Frozen solution. |
---|---|
Peso molecular | 59 kDa |
Especie | Human |
Recombinante | Recombinant |
Etiqueta de proteína | N-terminal 6His |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17300184
|
Eurofins DiscoverX
14-442 |
10 μg |
525.15€
10 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
17310184
|
Eurofins DiscoverX
14-442M |
250 μg |
4008.15€
250 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
The lck protooncogene is a member of the src-related gene family of non-receptor tyrosine kinases. It is uniquely expressed only in T-lymphocytes and NK-cells. The p56lck kinase (LCK) is a 56 KD polypeptide that is associated with a cytoplasmic face of the plasma membrane. It functions as a critical signalling model in IL-2 signal transduction leading to T-lymphocyte activation. Specifically, it has been shown that a stable complex exists between IL-2 receptors and p56lck and that treatment of T-lymphocytes with IL-2 promotes p56lck kinase activity and rapid phosphorylation of cellular substrates. In addition, p56lck has been found to function as a signal transducer for antigen activated CD4 and CG8 T-cell receptors and plays a role in thymic maturation of certain T-cells. The CD45 antigen, a receptor/protein tyrosine phosphatase of T-cells, also interacts with p56lck.
Swiss Prot P06239
Especificaciones
mg/mL of enzyme in 50 mM Tris/HCl, pH 7.5, 0.1 mM EGTA, 150 mM NaCl, 1 mM benzamidine, 0.03% Brij-35, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 5% glycerol. Frozen solution. | |
Human | |
N-terminal 6His | |
Full-length | |
Lck, active |
59 kDa | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥73% |