Learn More
Invitrogen™ Mouse IL-13, AOF Recombinant Protein
E. coli Recombinant Protein
Marca: Invitrogen™ RP87749
Descripción
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.- Purity: >95% by SDS-Page
- Endotoxin Concentration: AX41 ≤EUs/μg
- Protein Length: 121 aa.
- Biological Activity: <10 ng/mL
- Molecular Weight: 12.3 kDa
- Animal Origin Free: Yes

Especificaciones
P20109 | |
0.1% TFA with no preservative | |
12.3 kDa | |
10 μg | |
-20°C | |
ALRH; ALRH antibody; BHR1; BOS_7773; il 13; Il13; Il-13; ILN; Interleukin; interleukin 13; Interleukin13; Interleukin-13; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 | |
Mouse | |
Recombinant | |
>95% |
Control, ELISA, Gel Shift, Western Blot | |
16163 | |
Mouse IL-13, AOF | |
E. coli | |
RUO | |
Il13 | |
Unconjugated | |
Lyophilized |
Sugerencias de productos
Customers who viewed this item also viewed.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.