missing translation for 'onlineSavingsMsg'
Learn More
N-(2-Methoxy-5-methylphenyl)benzenesulfonamide, 97%, Thermo Scientific™ Product Code.: 15482336

N-(2-Methoxy-5-methylphenyl)benzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15482336
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15482336

Brand: Thermo Scientific Alfa Aesar H55897.03

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6964-02-9
MDL Number MFCD00085470

Specifications

Molecular Formula C8H18O2
Quantity 1 g
Synonym 3-methyl-2,4-heptanediol, 3-methylheptandiol-2.4
InChI Key ZMWAURRHTDCQCX-UHFFFAOYNA-N
SMILES CCCC(C(C)C(C)O)O
IUPAC Name 3-methylheptane-2,4-diol
Molecular Weight (g/mol) 146.23
PubChem CID 248872
Formula Weight 277.34
Percent Purity 97%
Chemical Name or Material N-(2-Methoxy-5-methylphenyl)benzenesulfonamide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberBL080301 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-(2-Methoxy-5-methylphenyl)benzenesulfonamide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.