Learn More
N-Methyl-3-(1-methyl-1H-pyrazol-3-yl)benzylamine, 90%, Thermo Scientific™ Product Code.: 10453204
N-Methyl-3-(1-methyl-1H-pyrazol-3-yl)benzylamine, 90%, Thermo Scientific™
Product Code. 10453204
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
Product Code. 10453204
Brand: Thermo Scientific Maybridge CC60646CB
Request Bulk or Custom Format
This item is not returnable. View return policy
Chemical Identifiers
CAS 910037-12-6
Molecular Formula C12H15N3
Molecular Weight (g/mol) 201.273
MDL Number MFCD09065011
InChI Key QTPPVJGJXALONT-UHFFFAOYSA-N
Synonym n-methyl-3-1-methyl-1h-pyrazol-3-yl benzylamine, methyl 3-1-methylpyrazol-3-yl phenyl methyl amine, n-methyl-1-3-1-methyl-1h-pyrazol-3-yl phenyl methanamine
PubChem CID 24229634
IUPAC Name N-methyl-1-[3-(1-methylpyrazol-3-yl)phenyl]methanamine
SMILES CNCC1=CC=CC(=C1)C2=NN(C=C2)C
Specifications
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 201.27
Percent Purity 90%
Chemical Name or Material N-methyl-3-(1-methyl-1H-pyrazol-3-yl)benzylamine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-Methyl-3-(1-methyl-1H-pyrazol-3-yl)benzylamine, 90%, Thermo Scientific™ >
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.