missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-N-(4-pyridin-4-ylbenzyl)amine, 90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 198.27 |
Percent Purity | 90% |
Chemical Name or Material | N-Methyl-N-(4-pyridin-4-ylbenzyl)amine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-Methyl-N-(4-pyridin-4-ylbenzyl)amine, 90%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction