missing translation for 'onlineSavingsMsg'
Learn More
N,N'-Diethyloxamide, 98%, Thermo Scientific™ Product Code.: 11481187

N,N'-Diethyloxamide, 98%, Thermo Scientific™

Product Code. 11481187
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11481187

Brand: Thermo Scientific Alfa Aesar B24167.14

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-84-9
Molecular Formula C6H12N2O2
Molecular Weight (g/mol) 144.17
MDL Number MFCD00043650
InChI Key FFYAVOJIYAAUNX-UHFFFAOYSA-N
Synonym nn'-diethyloxamide, ethanediamide, n,n'-diethyl, n,n'-diethyloxalamide, n,n-diethyloxamide, n,n'-diethylethanediamide, oxamide, n,n'-diethyl, acmc-1bc1a, n,n'-diethyl-oxalamide, n-ethyl-n'-ethylethane-1,2-diamide, n 1 ,n 2-diethylethanediamideShow MoreShow Less
PubChem CID 69210
SMILES CCNC(=O)C(=O)NCC

Specifications

Melting Point 178°C to 182°C
Odor Odorless
Quantity 25 g
IUPAC Name N,N'-diethylethanediamide
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material N,N'-Diethyloxamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N'-Diethyloxamide, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.