missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nestin Monoclonal Antibody (1A9G12), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA517136
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA5-17136 targets NES in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17136 immunogen is purified recombinant fragment of human NES (amino acids: 419-588 ) expressed in E. Coli. MA5-17136 detects NES which has a predicted molecular weight of approximately 177kDa.
Nestin is a Class VI intermediate filament abundantly produced in the developing central nervous system (CNS) and in early embryonic neuroepithelial stem cells. Nestin has been widely used as a predominant marker for stem/progenitor cells, glioma cells, and tumor endothelial cells. Also, Nestin is a superior angiogenic marker to evaluate neovascularity of endothelial cells in tumors. Highly specific antibodies to human Nestin can aid in characterizing progenitor cells differentiating into distinct lineages, in enhancing the therapeutic potential of human neural stem/progenitor cells in the treatment of CNS diseases or injury, and in identifying neuroepithelial tumor cells. Nestin expression has been observed in pancreatic hematopoietic stem cell populations, and is considered a novel angiogenesis marker of proliferating endothelial cells in colorectal cancer tissue.
Especificaciones
Nestin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P48681 | |
Nes | |
Purified recombinant fragment of human NES (amino acids: 419-588 ) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgM |
ELISA, Flow Cytometry, Western Blot | |
1A9G12 | |
Unconjugated | |
Nes | |
AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; Nestin | |
Mouse | |
Protein G | |
RUO | |
10763 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Nestin Monoclonal Antibody (1A9G12), Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido