missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMNAT1 Monoclonal Antibody (OTI1G4), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA525369
373.35 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT (EC 2.7.7.1) is a central enzyme in NAD biosynthesis, catalyzing the condensation of nicotinamide mononucleotide (NMN) or nicotinic acid mononucleotide (NaMN) with the AMP moiety of ATP to form NAD or NaAD.The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT (EC 2.7.7.1) is a central enzyme in NAD biosynthesis, catalyzing the condensation of nicotinamide mononucleotide (NMN) or nicotinic acid mononucleotide (NaMN) with the AMP moiety of ATP to form NAD or NaAD (Zhang et al., 2003 [PubMed 12574164]).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
Especificaciones
NMNAT1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9HAN9 | |
NMNAT1 | |
Full length human recombinant protein of NMNAT1 produced in HEK293T cell | |
100μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1G4 | |
Unconjugated | |
NMNAT1 | |
2610529L11Rik; 5730441G13Rik; D4Cole1e; deamido-NAD(+) diphosphorylase; deamido-NAD(+) pyrophosphorylase; EC 2.7.7.1; EC 2.7.7.18; id:ibd5068; im:7144541; LCA9; NAD(+) diphosphorylase; NAD(+) pyrophosphorylase; naMN adenylyltransferase 1; nicotinamide mononucleotide adenylyl transferase; nicotinamide mononucleotide adenylyl transferase 1; nicotinamide mononucleotide adenylyltransferase 1; nicotinamide mononucleotide adenylyltransferase 1-like protein; nicotinamide nucleotide adenylyltransferase 1; nicotinamide/nicotinate mononucleotide adenylyltransferase 1; nicotinamide/nicotinic acid mononucleotide adenylyltransferase 1; nicotinamide-nucleotide adenylyltransferase 1; Nicotinate-nucleotide adenylyltransferase 1; NMN adenylyltransferase 1; NMN/NaMN adenylyltransferase 1; nmnat; NMNAT1; NMNATPNAT1; PNAT1; PNAT-1; pyridine nucleotide adenylyltransferase 1; zgc:110243 | |
Mouse | |
Affinity Chromatography | |
RUO | |
64802 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
NMNAT1 Monoclonal Antibody (OTI1G4), Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido