missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otelixizumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Marca: Invitrogen MA541836
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Otelixizumab is a monoclonal antibody that binds to a receptor found on all T cells called CD3, which is involved in normal T cell signaling. Otelixizumab is believed to inhibit the function of autoreactive T cells, which are important in propagating autoimmune diseases, while inducing regulatory T cell pathways that promote immunological tolerance and inhibit autoreactive disease activity. Tolerx is developing otelixizumab to treat patients with type 1 diabetes, psoriasis and other autoimmune diseases such as rheumatoid arthritis.
Especificaciones
Otelixizumab | |
Recombinant Monoclonal | |
Unconjugated | |
ChAglyCD | |
Human CD3E | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
3.95 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 λ |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Otelixizumab Recombinant Human Monoclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido