missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PASD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA531463
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: PC-3, SK-N-SH. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that is thought to function as a transcription factor. The protein is a cancer-associated antigen that can stimulate autologous T-cell responses, and it is therefore considered to be a potential immunotherapeutic target for the treatment of various hematopoietic malignancies, including diffuse large B-cell lymphoma.
Especificaciones
PASD1 | |
Polyclonal | |
Unconjugated | |
PASD1 | |
Cancer/testis antigen 63; circadian clock protein PASD1; CT63; Gm1141; OXTES1; OX-TES-1; PAS domain containing 1; PAS domain-containing protein 1; PASD1; predicted gene 1141 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
139135 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IV76 | |
PASD1 | |
Recombinant protein fragment corresponding to a region within amino acids 503 and 721 of Human PASD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
PASD1 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido