missing translation for 'onlineSavingsMsg'
Learn More
(R)-(-)-1-Boc-3-hydroxypyrrolidine, 98%, Thermo Scientific Chemicals Product Code.: 15423727

(R)-(-)-1-Boc-3-hydroxypyrrolidine, 98%, Thermo Scientific Chemicals

Product Code. 15423727
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15423727

Brand: Thermo Scientific Alfa Aesar H27590.MD

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109431-87-0
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.24
MDL Number MFCD01317838
InChI Key APCBTRDHCDOPNY-UHFFFAOYNA-N
Synonym r-1-boc-3-hydroxypyrrolidine, r-1-n-boc-3-hydroxypyrrolidine, r---n-boc-3-pyrrolidinol, r-n-tert-butoxycarbonyl-3-hydroxypyrrolidine, tert-butyl 3r-3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-pyrrolidinol, r-bochp, r-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-hydropyrrolidine, r---boc-3-pyrrolidinol
PubChem CID 6544479
IUPAC Name tert-butyl (3R)-3-hydroxypyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(O)C1

Specifications

Melting Point 62°C to 65°C
Quantity 250 mg
Formula Weight 187.24
Percent Purity 98%
Chemical Name or Material (R)-(-)-1-Boc-3-hydroxypyrrolidine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberI23T036 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27590.03,H27590.MD,H27590.06
Lot Number10138239 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27590.03,H27590.MD,H27590.06
Lot NumberA08T023 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27590.03,H27590.MD,H27590.06
Lot NumberH12T016 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27590.03,H27590.MD,H27590.06
Lot Number10138261 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27590.03,H27590.MD,H27590.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-(-)-1-Boc-3-hydroxypyrrolidine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.