Learn More
Ractopamine Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marca: Invitrogen PA175375
Descripción
PA1-75375 specifically detects Ractopamine. PA1-75375 has been successfully used in ELISA applications. The PA1-75375 immunogen is Ractopamine(10)-BSA. Recommended Working Concentration: 2.5 μg/mL; Sensitivity: 10 ng/mL Ractopamine produces 72% inhibition in a competitive ELISA employing Ractopamine polyclonal antibody.
Ractopamine is a phenolethanolamine b-adrenoceptor agonist that is used for the improvement of weight gain, carcass leanness and feed efficiency in pigs. It has been known for years that ractopamine enhances growth in meat animals by repartitioning nutrients. This means that the nutrients that the animal consumes in its feed are shifted away from being stored as fat and are instead used in lean tissue production. This shift occurs as the animal reaches the top of its growth curve and lean tissue production drops off and fat deposition increases. Fat levels also appear to decrease, due to an increase in lypolysis (fat breakdown). Using the ractopamine monoclonal antibody, it is possible to detect ractopamine and other agonists in urine, muscle and liver both rapidly and with accuracy.
Especificaciones
Ractopamine | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Sheep | |
purified | |
RUO | |
Chemical | |
Antibody | |
IgG |
ELISA | |
Unconjugated | |
C18H23NO3 | |
Ractopamine(10)-BSA | |
1 mg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |