Learn More
TOM1L2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA554753
341.35 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
Immunogen sequence: LLHVNDDLNN VFLRYERFER YRSGRSVQNA SNGVLNEVTE DNLIDLGPGS PAVVSPMVGN TAPPSSLSSQ LAGLDLGTES VSGTLSSLQQ CNPRDGF Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
This gene belongs to a small gene family whose members have an N-terminal VHS domain followed by a GAT domain; domains which typically participate in vesicular trafficking. The canonical protein encoded by this gene also has a C-terminal clathrin binding motif. This protein has been shown to interact with Tollip, clathrin and ubiquitin and is thought to play a role in endosomal sorting. This gene resides in the 3.7 Mb deletion of chromosome region 17p11.2 that is associated with Smith-Magenis syndrome. Alternative splicing results in multiple transcript variants encoding distinct proteins.
Especificaciones
TOM1L2 | |
Polyclonal | |
Unconjugated | |
TOM1L2 | |
2900016I08Rik; A730055F12Rik; AU042072; FLJ32746; myb1-like protein 2; Srebf1; target of myb1 like 2 membrane trafficking protein; target of myb1-like 2; target of myb1-like 2 (chicken); Target of Myb-like protein 2; Tom1l2; TOM1-like protein 2; UNQ3124 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
146691 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZVM7 | |
TOM1L2 | |
Recombinant protein corresponding to Human TOM1L2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Research Use Only