missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tositumomab Recombinant Mouse Monoclonal Antibody, Invitrogen™
Mouse Recombinant Monoclonal Antibody
Marca: Invitrogen MA541763
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Murine IgG2a lambda monoclonal antibody against CD20 antigen (2 heavy chains of 451 residues, 2 lambda chains of 220 residues). It is produced in an antibiotic-free culture of mammalian cells. It can be covalently linked to Iodine 131 (a radioactive isotope of iodine). Binds to the CD20 antigen which is found on mature B lymphocytes. The antibody binding appears to induce apoptosis, complement-dependent cytotoxicity and cell death through ionizing radiation.
Especificaciones
Tositumomab | |
Recombinant Monoclonal | |
Unconjugated | |
CD20; MS4A1 | |
Human MS4A1/CD20 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG2a λ |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Tositumomab Recombinant Mouse Monoclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido