missing translation for 'onlineSavingsMsg'
Learn More
4-(2-Bromoethyl)-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific™ Product Code.: 10432613

4-(2-Bromoethyl)-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10432613
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10432613

Brand: Thermo Scientific Maybridge MO01241DA

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Chemical Identifiers

CAS 83467-28-1
Molecular Formula C7H11BrN2
Molecular Weight (g/mol) 203.08
MDL Number MFCD00480987
InChI Key HGGLMDBNLJUEKA-UHFFFAOYSA-N
Synonym 4-2-bromoethyl-3,5-dimethyl-1h-pyrazole, 4-2-bromo-ethyl-3,5-dimethyl-1h-pyrazole, 3,5-dimethyl-4-2-bromoethyl-1h-pyrazole, 4-2-bromoethyl-3,5-dimethyl-1h-pyrazol, 1h-pyrazole, 4-2-bromoethyl-3,5-dimethyl, 4-2-bromoethyl-3,5-dimethylpyrazole, akos pao-0750, pubchem12761, timtec-bb sbb010155, aronis23735Show MoreShow Less
PubChem CID 572879
IUPAC Name 4-(2-bromoethyl)-3,5-dimethyl-1H-pyrazole
SMILES CC1=C(CCBr)C(C)=NN1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 203.08
Percent Purity 97%
Chemical Name or Material 4-(2-Bromoethyl)-3, 5-dimethyl-1H-pyrazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(2-Bromoethyl)-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.