Learn More
adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human APPL1 protein.
Marca: Abnova H00026060-D01P.100ug
Descripción
The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signalling and insulin signalling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus. [provided by RefSeq
Sequence: MPGIDKLPIEETLEDSPQTRSLLGVFEEDATAISNYMNQLYQAMHRIYDAQNELSAATHLTSKLLKEYEKQRFPLGGDDEVMSSTLQQFSKVIDELSSCHAVLSTQLADAMMFPITQFKERDLKEILTLKEVFQIASNDHDAAINRYSRLSKKRENDKVKYEVTEDVYTSRKKQHQTMMHYFCALNTLQYKKKIALLEPLLGYMQAQISFFKMGSENLNEQLEEFLANIGTSVQNVRREMDSDIETMQQTIEDLEVASDPLYVPDPDPTKFPVNRNLTRKAGYLNARNKTGLVSSTWDRQFYFTQGGNLMSQARGDVAGGLAMDIDNCSVMAVDCEDRRYCFQITSFDGKKSSILQAESKKDHEEWICTINNISKQIYLSENPEETAARVNQSALEAVTPSPSFQQRHESLRPAAGQSRPPTARTSSSGSLGSESTNLAALSLDSLVAPDTPIQFDIISPVCEDQPGQAKAFGQGGRRTNPFGESGGSTKSETEDSILHQLFIVRFLGSMEVKSDDHPDVVYETMRQILAARAIHNIFRMTESHLLVTCDCLKLIDPQTQVTRLTFPLPCVVLYATHQENKRLFGFVLRTSSGRSESNLSSVCYIFESNNEGEKICDSVGLAKQIALHAELDRRASEKQKEIERVKEKQQKELNKQKQIEKDLEEQSRLIAASSRPNQASSEGQFVVLSSSQSEESDLGEGGKKRESEAEspecificaciones
adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human APPL1 protein. | |
APPL1 | |
APPL/DIP13alpha | |
Rabbit | |
Affinity Purified | |
RUO | |
26060 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
In situ PLA, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_012096 | |
APPL1 | |
APPL1 (NP_036228.1, 1 a.a. ∼ 709 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.