Learn More
CD55 Mouse anti-Human, PE-Cy5, Clone: IA10, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 555695
Descripción
Reacts with CD55, decay-accelerating factor (DAF), a glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein of approximately 70 kDa, expressed on hematopoietic cells. It has been suggested that the role of DAF is to protect cells from damage by autologous complement preventingthe amplification steps of the complement cascade by interfering with the assembly of the C3-convertases, C4b2a and C3bBb, and the C5-convertases, C4b2a3b and C3bBb3b.

Especificaciones
CD55 | |
Monoclonal | |
PE-Cyanine5 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
IA10 | |
DAF | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG2a κ |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.