missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium tuberculosis CFP-10 Rabbit anti-Bacteria, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA119445
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody recognizes CFP10 of Mycobacterium tuberculosis H37Rv and Mycobacterium bovis; it reacts also with recombit antigen produced in Escherichia coli.
Tuberculosis is a chronic bacterial infection which remains one of the leading killers of young adults world wide. Mycobacterium tuberculosis enters the alveoli by airborne transmission. They are engulfed by alveolar macrophages but resist destruction. Although the exact mechanism is not known, it is suspected that that Mycrobacterium tuberculosis survives by preventing the fusion of the phagosome and lysosome. As the bacterium accumulates it forms a tubercle, spreads to the lymph nodes and becomes a systemic infection. Although treatments for tuberculosis do exist, new strains of Mycobacterium tuberculosis that appear to have multiple-drug resistance (MDR) pose a considerable threat to public health.
Especificaciones
Mycobacterium tuberculosis CFP-10 | |
Polyclonal | |
Unconjugated | |
M. tuberculosis; myobacter | |
CFP10 (Rv3874) of Mycobacterium tuberculosis. | |
100 μg | |
Primary | |
4° C, do not freeze | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Mycobacterium tuberculosis CFP-10 Rabbit anti-Bacteria, Polyclonal, Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido