missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Coagulation Factor III/Tissue Factor Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
Marca: Novus Biologicals™ NBP2-79650-15PLATES
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This solid phase sandwich ELISA utilizes an antibody specific for Rat Coagulation Factor III/Tissue Factor.
Especificaciones
23.45 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes an antibody specific for Rat Coagulation Factor III/Tissue Factor. | |
Sandwich ELISA | |
2152 | |
Rabbit/Mouse | |
15 Plates | |
Cancer | |
Coagulation Factor III/Tissue Factor |
23.45-1500 pg/mL | |
HRP | |
Antibody Pairs | |
Rat | |
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
F3 | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended. |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Novus Biologicals™ Coagulation Factor III/Tissue Factor Antibody Pair [HRP] > 15 Plates; HRP
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido