Learn More
CRF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA5111543
Descripción
Immunogen sequence: CRALLSRGPV PGARQAPQHP QPLDFFQPPP QSEQPQQPQA RPVLLRMGE
Corticotropin Releasing Factor (CRF), a 41 amino acids neuropeptide, belongs to a family of structurally related peptides including urocortin, sauvagine and urotensin I. CRF is critical in the regulation of the pituitary-adrenal gland axis, and in endocrine, autonomic and behavioral responses to stress. CRF is widely distributed in the brain and peripheral nervous system with the highest levels in the hypothalamic paraventricular nucleus.
Especificaciones
CRF | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P06850, Q13324 | |
CRH, Crhr2 | |
Recombinant Protein corresonding to Human CRF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
Crhr2 | |
Corticoliberin; Corticotropin-releasing factor; Corticotropin-releasing factor receptor 2; Corticotropin-releasing hormone; Corticotropin-releasing hormone receptor 2; CRF; CRF2; CRF2R; Crfr2; CRF-R2; CRF-R-2; CRFR2alpha; CRFR2beta; CRF-RB; CRH; CRH1; CRH2R; Crhr2; CRH-R-2; Gm1347; HM-CRF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1392, 1395 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Sugerencias de productos
Customers who viewed this item also viewed.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.